Learn More
Medchemexpress LLC 5α-Cholestan-3-one | 566-88-1 | MFCD00065901 | 99.3% | 386.65 g/mol | C27H46O | 250 MG
SDP
Catalog No. 5000241706
Encompass_Preferred
5α-Cholestan-3-one is an endogenous steroid metabolite supplied as a high-purity analytical standard for research and analytical applications. It is a white to off-white solid with molecular formula C27H46O, molecular weight 386.65 g/mol, and a reported purity of 99.33%.
  • High purity (99.33%) for analytical accuracy and reproducibility.
  • Solid form suitable for convenient storage and handling.
  • Available in small-scale quantities for method development and validation.
  • Appropriate for biochemical and metabolic studies as a reference standard.
  • Comes with datasheet and certificate of analysis for lot traceability.
Catalog No. 50-002-41706 Supplier Medchemexpress LLC Supplier No. HY107826250MG
5α-Cholestan-3-one is an endogenous steroid metabolite supplied as a high-purity analytical standard for research and analytical applications. It is a white to off-white solid with molecular formula C27H46O, molecular weight 386.65 g/mol, and a reported purity of 99.33%.
  • High purity (99.33%) for analytical accuracy and reproducibility.
  • Solid form suitable for convenient storage and handling.
  • Available in small-scale quantities for method development and validation.
  • Appropriate for biochemical and metabolic studies as a reference standard.
  • Comes with datasheet and certificate of analysis for lot traceability.