Learn More
Medchemexpress LLC 5α-cholestane-2,2,3,3,4,4-d6 | 358731-18-7 | MFCD01861631 | >98.0% | C27H42D6 | 10MG
SDP
Catalog No. 5000261925
Encompass_Preferred
5α-Cholestane-d6 is the deuterium-labeled (d6) analog of 5α-cholestane supplied as an isotope-labeled reference standard for analytical and synthetic applications. It is intended for use as an internal standard in mass spectrometry, for tracer studies, and for synthetic chemistry where a high-purity, isotopically enriched cholestane derivative is required.
  • Deuterium-labeled standard (d6) for isotope dilution and tracing.
  • High chemical purity (>98.0%) suitable for analytical use.
  • Isotopic enrichment approximately 95 atom % D for clear mass shift.
  • Molecular formula C27H42D6; molecular weight 378.71.
  • Provided in small quantities appropriate for reference and analytical workflows.
Catalog No. 50-002-61925 Supplier Medchemexpress LLC Supplier No. HY133970S10MG
5α-Cholestane-d6 is the deuterium-labeled (d6) analog of 5α-cholestane supplied as an isotope-labeled reference standard for analytical and synthetic applications. It is intended for use as an internal standard in mass spectrometry, for tracer studies, and for synthetic chemistry where a high-purity, isotopically enriched cholestane derivative is required.
  • Deuterium-labeled standard (d6) for isotope dilution and tracing.
  • High chemical purity (>98.0%) suitable for analytical use.
  • Isotopic enrichment approximately 95 atom % D for clear mass shift.
  • Molecular formula C27H42D6; molecular weight 378.71.
  • Provided in small quantities appropriate for reference and analytical workflows.