Learn More
Medchemexpress LLC 5-Cholesten-24(RS)-methyl-3β-ol-d3 | 23929-42-2 | 99.2% | 403.70 | 1 MG
SDP
Catalog No. 5000363628
Encompass_Preferred
5-Cholesten-24(RS)-methyl-3β-ol-d3 is a deuterium labeled compound intended for research use. It can serve as a tracer or an internal standard for quantitative analysis. Deuteration is noted for its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development.
Catalog No. 50-003-63628 Supplier Medchemexpress LLC Supplier No. HY146627S1MG
Add to Cart
Add to Cart
5-Cholesten-24(RS)-methyl-3β-ol-d3 is a deuterium labeled compound intended for research use. It can serve as a tracer or an internal standard for quantitative analysis. Deuteration is noted for its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development.