Learn More
Biotium 5-CARBOXYRhodamine 6G, SE (5-CR 6G, SE)
Catalog No. 501964807
Encompass_Preferred
Amine-reactive form of 5-carboxyrhodamine 6G single isomer. Properties: Ex/ Em(MeOH) = 520/546 nm; Orange red solid moderately soluble in DMSO; Store at -20°C and protect from light; C31H29ClN3O7; MW: 555.6
Catalog No. 50-196-4807 Supplier Biotium Supplier No. 90032
Add to Cart
Edge
Add to Cart
Amine-reactive form of 5-carboxyrhodamine 6G single isomer. Properties: Ex/ Em(MeOH) = 520/546 nm; Orange red solid moderately soluble in DMSO; Store at -20°C and protect from light; C31H29ClN3O7; MW: 555.6