Learn More
Medchemexpress LLC 5-Cyano-2,3-di-(p-tolyl)tetrazolium chloride | 90217-02-0 | 98.1% | 5 MG
SDP
Catalog No. 5000410806
Encompass_Preferred
5-Cyano-2,3-di-(p-tolyl)tetrazolium chloride, also known as CTC, is a redox-sensitive red fluorescent dye. It is primarily used for detecting metabolic activity in microorganisms. The compound has an emission maximum at 602 nm.
  • Redox-sensitive fluorescent dye.
  • Detects metabolic activity.
  • Fluorescent, emitting at 602 nm.
  • Solid, white to off-white in color.
Catalog No. 50-004-10806 Supplier Medchemexpress LLC Supplier No. HYD00165MG
5-Cyano-2,3-di-(p-tolyl)tetrazolium chloride, also known as CTC, is a redox-sensitive red fluorescent dye. It is primarily used for detecting metabolic activity in microorganisms. The compound has an emission maximum at 602 nm.
  • Redox-sensitive fluorescent dye.
  • Detects metabolic activity.
  • Fluorescent, emitting at 602 nm.
  • Solid, white to off-white in color.