Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Cyano-2-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C32275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3227-5G 5 g
C32271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3227-5G Supplier TCI America Supplier No. C32275G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 468718-30-1
Molecular Formula C7H5BFNO2
Molecular Weight (g/mol) 164.93
MDL Number MFCD03095147
InChI Key ITCSMTHUTFTXLE-UHFFFAOYSA-N
PubChem CID 2783361
IUPAC Name (5-cyano-2-fluorophenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)C#N)F)(O)O

Specifications

Color White
Quantity 5 g
Synonym 5-cyano-2-fluorobenzeneboronic acid, 5-cyano-2-fluorophenyl boronic acid, 3-borono-4-fluorobenzonitrile, boronic acid, 5-cyano-2-fluorophenyl, 5-cyano-2-fluoro-phenyl boronic acid
Formula Weight 164.93
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.