Learn More
Medchemexpress LLC 5-cyclohexyl-3,3-dimethyl-1-[4-[2-(oxan-4-ylamino)ethylamino]phenyl]-2-oxo-1,4-benzodiazepine | 2376334-75-5 | 99.4% | 513.67 g/mol | C31H39N5O2 | 50MG
SDP
Catalog No. 5000238159
Encompass_Preferred
This small-molecule parathyroid hormone receptor 1 (PTHR1) antagonist is a potent, orally bioavailable compound (IC50 = 90 nM) intended for research use. It is supplied as a light yellow to yellow solid with high reported purity and defined physicochemical properties suitable for in vitro and in vivo preclinical studies.
  • Potent PTHR1 antagonist with IC50 of 90 nM.
  • Orally bioavailable in preclinical models.
  • High purity (99.4%) and solid form, light yellow to yellow.
  • Molecular formula C31H39N5O2 and molecular weight 513.67 g/mol.
  • Multiple pack sizes available to support assay scale-up.
  • Supplied for research use only.
Catalog No. 50-002-38159 Supplier Medchemexpress LLC Supplier No. HY12587950MG
This small-molecule parathyroid hormone receptor 1 (PTHR1) antagonist is a potent, orally bioavailable compound (IC50 = 90 nM) intended for research use. It is supplied as a light yellow to yellow solid with high reported purity and defined physicochemical properties suitable for in vitro and in vivo preclinical studies.
  • Potent PTHR1 antagonist with IC50 of 90 nM.
  • Orally bioavailable in preclinical models.
  • High purity (99.4%) and solid form, light yellow to yellow.
  • Molecular formula C31H39N5O2 and molecular weight 513.67 g/mol.
  • Multiple pack sizes available to support assay scale-up.
  • Supplied for research use only.