Learn More
Selleck Chemical LLC 2-Deoxyadenosine 5-monophosphate S3248-5mg
SDP
Catalog No. 503108136
Encompass_Preferred
2 -Deoxyadenosine 5 -monophosphate (5 -dAMP) an endogenous metabolite is only photosensitized degraded under acidic conditions
Catalog No. 50-310-8136 Supplier Selleck Chemical LLC Supplier No. S32485MG
Add to Cart
Add to Cart
2 -Deoxyadenosine 5 -monophosphate (5 -dAMP) an endogenous metabolite is only photosensitized degraded under acidic conditions