Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Dbco-Peg4-Dutp, 10Mm Aqueous Solution

Catalog No. 80970550UL Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-970-550UL 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 80-970-550UL Supplier Sigma Aldrich Supplier No. 80970550UL
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

5-DBCO-PEG4-dUTP is recommended for two-step labeling of DNA/cDNA e.g. by PCR with Taq polymerase. It is enzymatically incorporated into DNA/cDNA as a substitute for its natural counterpart dTTP. The resulting DBCO-functionalized DNA/cDNA can be labeled with azide-labeled molecules using copper-free click chemistry. This enables experiments such as introduction of a biotin group for purification tasks, introduction of a fluorescent group for detection, or crosslinking the DNA to azide-functionalized biomolecules such as proteins.

Specifications

CAS 2101-88-4
Molecular Formula C6H4BrN3
Linear Formula C6H4BrN3
MDL Number MFCD09281823
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.