Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-dehydroepisterol | 23582-83-4 | 92.4% | 396.6 g/mol | C28H44O | 5 MG
SDP

Catalog No. 5000262232
Encompass_Preferred

5-dehydroepisterol is an episterol derivative and an intermediate in steroid biosynthesis. It is used as a research reagent in studies of sterol metabolism and antifungal activity. The compound is supplied with a reported purity of 92.36%.

  • Episterol derivative and intermediate in steroid biosynthesis.
  • Reported antifungal activity.
  • Purity 92.36% as supplied.
  • Molecular formula C28H44O; molecular weight 396.6 g/mol.
  • Suitable for biochemical and sterol metabolism research applications.

Catalog No. 50-002-62232 Supplier Medchemexpress LLC Supplier No. HY1307035MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-dehydroepisterol is an episterol derivative and an intermediate in steroid biosynthesis. It is used as a research reagent in studies of sterol metabolism and antifungal activity. The compound is supplied with a reported purity of 92.36%.

  • Episterol derivative and intermediate in steroid biosynthesis.
  • Reported antifungal activity.
  • Purity 92.36% as supplied.
  • Molecular formula C28H44O; molecular weight 396.6 g/mol.
  • Suitable for biochemical and sterol metabolism research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.