Learn More
Medchemexpress LLC 5-dehydroepisterol | 23582-83-4 | MFCD32669850 | 92.4% | C28H44O | 10MG
SDP
Catalog No. 5000261206
Encompass_Preferred
5-Dehydroepisterol is an episterol-derived sterol intermediate used in steroid biosynthesis and biochemical research. It is provided as a solid (white to yellow) with molecular formula C28H44O and molecular weight ~396.65 g/mol. Intended for research and analytical applications; follow recommended storage and handling to preserve sample integrity.
  • Sterol intermediate used in steroid biosynthesis studies.
  • Supplied as a solid, white to yellow, for straightforward handling.
  • High purity suitable for analytical and research applications.
  • Stable when stored under recommended low-temperature, light-protected conditions.
  • Clear handling and storage guidance to maintain sample integrity.
Catalog No. 50-002-61206 Supplier Medchemexpress LLC Supplier No. HY13070310MG
5-Dehydroepisterol is an episterol-derived sterol intermediate used in steroid biosynthesis and biochemical research. It is provided as a solid (white to yellow) with molecular formula C28H44O and molecular weight ~396.65 g/mol. Intended for research and analytical applications; follow recommended storage and handling to preserve sample integrity.
  • Sterol intermediate used in steroid biosynthesis studies.
  • Supplied as a solid, white to yellow, for straightforward handling.
  • High purity suitable for analytical and research applications.
  • Stable when stored under recommended low-temperature, light-protected conditions.
  • Clear handling and storage guidance to maintain sample integrity.