Learn More
Medchemexpress LLC 5 -Deoxy-2 3 -di-O- 25g
SDP
Catalog No. 5000206142
Encompass_Preferred
5 -Deoxy-2 3 -di-O-acetyl-5-fluorocytidine (2 3 -Di-O-acetyl-5 -deoxy-5-fluorocytidine) is an antiviral compound that inhibits HBV reverse transcriptase activity 5 -Deoxy-2 3 -di-O-acetyl-5-fluorocytidine shows potential to control HBV replication in vitro 5 -Deoxy-2 3 -di-O-acetyl-5-fluorocytidine as a nucleoside analog may provide a new option for the suppression of chronic hepatitis B[1]
Catalog No. 50-002-06142 Supplier Medchemexpress LLC Supplier No. 25GHYW105601
5 -Deoxy-2 3 -di-O-acetyl-5-fluorocytidine (2 3 -Di-O-acetyl-5 -deoxy-5-fluorocytidine) is an antiviral compound that inhibits HBV reverse transcriptase activity 5 -Deoxy-2 3 -di-O-acetyl-5-fluorocytidine shows potential to control HBV replication in vitro 5 -Deoxy-2 3 -di-O-acetyl-5-fluorocytidine as a nucleoside analog may provide a new option for the suppression of chronic hepatitis B[1]