Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5′-deoxy-5-fluoro-N4-(isopentyloxycarbonyl)cytidine | 162204-30-0 | MFCD00930630 | >95.0% | 359.35 g/mol | C15H22FN3O6 | 5 MG
SDP

Catalog No. 5000222953
Encompass_Preferred

5′-Deoxy-5-fluoro-N4-(isopentyloxycarbonyl)cytidine is an identified synthetic impurity produced in the synthesis of the anticancer prodrug capecitabine. Supplied for research use only, it is intended for analytical reference and impurity profiling and has documented physicochemical, storage, and in vitro activity data.

  • impurity reference for capecitabine synthesis
  • white to off-white solid appearance
  • CAS 162204-30-0
  • molecular formula C15H22FN3O6; molecular weight 359.35 g/mol
  • soluble in methanol at ≥ 50 mg/mL
  • storage: powder at -20°C (long term); solutions at -80°C (recommended)
  • available in small milligram pack sizes for analytical work
  • batch analytical data and documentation (COA, HNMR, LCMS) available

Catalog No. 50-002-22953 Supplier Medchemexpress LLC Supplier No. HY1345545MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5′-Deoxy-5-fluoro-N4-(isopentyloxycarbonyl)cytidine is an identified synthetic impurity produced in the synthesis of the anticancer prodrug capecitabine. Supplied for research use only, it is intended for analytical reference and impurity profiling and has documented physicochemical, storage, and in vitro activity data.

  • impurity reference for capecitabine synthesis
  • white to off-white solid appearance
  • CAS 162204-30-0
  • molecular formula C15H22FN3O6; molecular weight 359.35 g/mol
  • soluble in methanol at ≥ 50 mg/mL
  • storage: powder at -20°C (long term); solutions at -80°C (recommended)
  • available in small milligram pack sizes for analytical work
  • batch analytical data and documentation (COA, HNMR, LCMS) available

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.