Learn More
Medchemexpress LLC 5′-deoxy-5-fluoro-N4-(isopentyloxycarbonyl)cytidine | 162204-30-0 | MFCD00930630 | C15H22FN3O6 | 10MG
SDP
Catalog No. 5000259223
Encompass_Preferred
5′-Deoxy-5-fluoro-N4-(isopentyloxycarbonyl)cytidine is a small-molecule impurity formed during the synthesis of the anticancer prodrug capecitabine. It is provided as a research and analytical reference for laboratories performing impurity profiling, method development, and quality-control testing of synthetic processes.
  • Provided as an analytical reference standard for impurity profiling.
  • Suitable for HPLC, LC-MS, and method validation.
  • Supplied in small milligram quantities for laboratory use.
  • Molecular formula C15H22FN3O6 and molecular weight 359.35 g·mol-1.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-59223 Supplier Medchemexpress LLC Supplier No. HY13455410MG
5′-Deoxy-5-fluoro-N4-(isopentyloxycarbonyl)cytidine is a small-molecule impurity formed during the synthesis of the anticancer prodrug capecitabine. It is provided as a research and analytical reference for laboratories performing impurity profiling, method development, and quality-control testing of synthetic processes.
  • Provided as an analytical reference standard for impurity profiling.
  • Suitable for HPLC, LC-MS, and method validation.
  • Supplied in small milligram quantities for laboratory use.
  • Molecular formula C15H22FN3O6 and molecular weight 359.35 g·mol-1.
  • Intended for research use only; not for human therapeutic use.