Learn More
Medchemexpress LLC 5'-Deoxy-5-fluorocytidine | 66335-38-4 | 99.9% | 245.21 | 25 G
SDP
Catalog No. 5000454115
Encompass_Preferred
5'-Deoxy-5-fluorocytidine is a cytidine analog and a metabolite of Capecitabine. It undergoes enzymatic conversions in the body, ultimately producing 5-fluorouracil, which exerts anti-tumor effects. This compound is used in research for various solid tumors.
  • Cytidine analog
  • Metabolite of Capecitabine
  • Converted to 5-fluorouracil for anti-tumor effects
  • Utilized in research for solid tumors, including colorectal cancer, non-small cell lung cancer, and breast cancer
  • Inhibits proliferation of chemotherapy-resistant A549 paraclone cells in vitro
Catalog No. 50-004-54115 Supplier Medchemexpress LLC Supplier No. 25GHYW009538
5'-Deoxy-5-fluorocytidine is a cytidine analog and a metabolite of Capecitabine. It undergoes enzymatic conversions in the body, ultimately producing 5-fluorouracil, which exerts anti-tumor effects. This compound is used in research for various solid tumors.
  • Cytidine analog
  • Metabolite of Capecitabine
  • Converted to 5-fluorouracil for anti-tumor effects
  • Utilized in research for solid tumors, including colorectal cancer, non-small cell lung cancer, and breast cancer
  • Inhibits proliferation of chemotherapy-resistant A549 paraclone cells in vitro