Learn More
Medchemexpress LLC 5'-Deoxy-5-fluorocytidine (Standard) | 66335-38-4 | 99.9% | 245.21 | 1 ML
SDP
Catalog No. 5000453367
Encompass_Preferred
5'-Deoxy-5-fluorocytidine is a cytidine analog and a metabolite of Capecitabine. It is converted from Capecitabine by carboxylesterase in the liver. This compound is deaminated by cytidine deaminase to generate 5'-deoxy-5-fluorouridine, which is then converted into 5-fluorouracil by thymidine phosphorylase in tumor tissues to exert anti-tumor effects. It is used in research for solid tumors such as colorectal cancer, non-small cell lung cancer, and breast cancer.
  • Significantly inhibits the proliferation of chemotherapy-resistant A549 paraclone cells.
  • Used in research for solid tumors such as colorectal cancer, non-small cell lung cancer, and breast cancer.
  • Cytidine analog and metabolite of capecitabine.
  • Converted to 5-fluorouracil by thymidine phosphorylase in tumor tissues.
Catalog No. 50-004-53367 Supplier Medchemexpress LLC Supplier No. HYW00953810MM/1ML
5'-Deoxy-5-fluorocytidine is a cytidine analog and a metabolite of Capecitabine. It is converted from Capecitabine by carboxylesterase in the liver. This compound is deaminated by cytidine deaminase to generate 5'-deoxy-5-fluorouridine, which is then converted into 5-fluorouracil by thymidine phosphorylase in tumor tissues to exert anti-tumor effects. It is used in research for solid tumors such as colorectal cancer, non-small cell lung cancer, and breast cancer.
  • Significantly inhibits the proliferation of chemotherapy-resistant A549 paraclone cells.
  • Used in research for solid tumors such as colorectal cancer, non-small cell lung cancer, and breast cancer.
  • Cytidine analog and metabolite of capecitabine.
  • Converted to 5-fluorouracil by thymidine phosphorylase in tumor tissues.