Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-Deoxy-5-fluorouridine 98.0+%, TCI America™
SDP

Catalog No. D35795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35795G 5 g
D35791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35795G Supplier TCI America Supplier No. D35795G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9-5-3094
Molecular Formula C9H11FN2O5
Molecular Weight (g/mol) 246.19
MDL Number MFCD00866530 MFCD00866530
InChI Key ZWAOHEXOSAUJHY-ZIYNGMLESA-N
PubChem CID 18343
ChEBI CHEBI:31521
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES C[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=C(F)C(=O)NC1=O

Specifications

Melting Point 189°C
Color White
Quantity 5 g
Synonym 5′-dFUrd, Doxifluridine, 1-(5-Deoxy-beta-D-ribofuranosyl)-5-fluorouracil
Formula Weight 246.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5′-Deoxy-5-fluorouridine
RTECSNumber YU7526000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.