Learn More
Medchemexpress LLC 5'-Deoxy-5'-iodouridine | 14259-58-6 | 95.0% | C9H11IN2O5 | 1 ML
SDP
Catalog No. 5000434540
Encompass_Preferred
5'-Deoxy-5'-iodouridine is a purine nucleoside analog. Purine nucleoside analogs have broad antitumor activity targeting indolent lymphoid malignancies. Anticancer mechanisms in this process rely on inhibition of DNA synthesis, induction of apoptosis, etc.
  • Purine nucleoside analog
  • Broad antitumor activity targeting indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
Catalog No. 50-004-34540 Supplier Medchemexpress LLC Supplier No. HY7764410MM/1ML
5'-Deoxy-5'-iodouridine is a purine nucleoside analog. Purine nucleoside analogs have broad antitumor activity targeting indolent lymphoid malignancies. Anticancer mechanisms in this process rely on inhibition of DNA synthesis, induction of apoptosis, etc.
  • Purine nucleoside analog
  • Broad antitumor activity targeting indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis