Learn More
Medchemexpress LLC 5'-Deoxy-5'-iodouridine | 14259-58-6 | 95.0% | C9H11IN2O5 | 100 MG
SDP
Catalog No. 5000435104
Encompass_Preferred
5'-Deoxy-5'-iodouridine is a purine nucleoside analog. Purine nucleoside analogs exhibit broad antitumor activity, specifically targeting indolent lymphoid malignancies. The anticancer mechanisms involved include the inhibition of DNA synthesis and the induction of apoptosis.
  • It is a purine nucleoside analog.
  • Demonstrates broad antitumor activity.
  • Targets indolent lymphoid malignancies.
  • Functions by inhibiting DNA synthesis.
  • Induces apoptosis.
Catalog No. 50-004-35104 Supplier Medchemexpress LLC Supplier No. HY77644100MG
5'-Deoxy-5'-iodouridine is a purine nucleoside analog. Purine nucleoside analogs exhibit broad antitumor activity, specifically targeting indolent lymphoid malignancies. The anticancer mechanisms involved include the inhibition of DNA synthesis and the induction of apoptosis.
  • It is a purine nucleoside analog.
  • Demonstrates broad antitumor activity.
  • Targets indolent lymphoid malignancies.
  • Functions by inhibiting DNA synthesis.
  • Induces apoptosis.