Learn More
Cayman Chemical 5-deoxy-D-Ribose, Purity: ≥95%, CAS: 13039-75-3, 100 mg
SDP
Catalog No. 502991306
Encompass_Preferred
A building block; has been used in the synthesis of carcinolytic compounds
Catalog No. 50-299-1306 Supplier Cayman Chemical Supplier No. 30103100 MG
A building block; has been used in the synthesis of carcinolytic compounds