Learn More
Cayman Chemical 5'-Deoxyadenosine, Purity: ≥95%, CAS: 4754-39-6, 100 mg
SDP
Catalog No. 502992563
Encompass_Preferred
An adenosine analog; an intermediate in the degradation of S-adenosyl-L-methionine; has been used in the study of enzyme kinetics, including those of phosphomethylpyrimidine synthase, glutamate mutase, and 5’-methylthioadenosine phosphorylase
Catalog No. 50-299-2563 Supplier Cayman Chemical Supplier No. 29619100 MG
An adenosine analog; an intermediate in the degradation of S-adenosyl-L-methionine; has been used in the study of enzyme kinetics, including those of phosphomethylpyrimidine synthase, glutamate mutase, and 5’-methylthioadenosine phosphorylase