Learn More
Cayman Chemical 5-DeoxyadenosIn 10mg
SDP
Catalog No. 502989519
Encompass_Preferred
An adenosine analog; an intermediate in the degradation of S-adenosyl-L-methionine; has been used in the study of enzyme kinetics, including those of phosphomethylpyrimidine synthase, glutamate mutase, and 5’-methylthioadenosine phosphorylase
Catalog No. 50-298-9519 Supplier Cayman Chemical Supplier No. 2961910 MG
An adenosine analog; an intermediate in the degradation of S-adenosyl-L-methionine; has been used in the study of enzyme kinetics, including those of phosphomethylpyrimidine synthase, glutamate mutase, and 5’-methylthioadenosine phosphorylase