Learn More
TARGETMOL CHEMICALS INC 5-DEOXYADENOSINE 25MG
SDP
Catalog No. 502579845
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 5'-Deoxyadenosine (5′-dAdo) is an oxidized nucleoside found in the urine of normal subjects. Oxidized nucleosides represent excellent biomarkers for determining the extent of damage in genetic material which has long been of interest in understanding the mechanism of aging neurodegenerative diseases and carcinogenesis. The normal form of deoxyadenosine used in DNA synthesis and repair is 2'-deoxyadenosine where the hydroxyl group (-OH) is at the 2' position of its ribose sugar moiety. 5'-deoxyadenosine has its hydroxyl group at the 5' position of the ribose sugar. purity: 99%
Catalog No. 50-257-9845 Supplier TARGETMOL CHEMICALS INC Supplier No. T497125MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 5'-Deoxyadenosine (5′-dAdo) is an oxidized nucleoside found in the urine of normal subjects. Oxidized nucleosides represent excellent biomarkers for determining the extent of damage in genetic material which has long been of interest in understanding the mechanism of aging neurodegenerative diseases and carcinogenesis. The normal form of deoxyadenosine used in DNA synthesis and repair is 2'-deoxyadenosine where the hydroxyl group (-OH) is at the 2' position of its ribose sugar moiety. 5'-deoxyadenosine has its hydroxyl group at the 5' position of the ribose sugar. purity: 99%