Learn More
Medchemexpress LLC 5'-Deoxyadenosine | 4754-39-6 | 99.9% | 251.24 | 10 MM * 1 ML
SDP
Catalog No. 5000435589
Encompass_Preferred
5'-Deoxyadenosine is an oxidized nucleoside found in the urine of normal subjects. It shows anti-orthopoxvirus activity.
  • Oxidized nucleoside
  • Found in the urine of normal subjects
  • Shows anti-orthopoxvirus activity
  • For research use only
  • Available in solid and solution forms
  • High purity (99.92%)
  • Endogenous metabolite
  • Human gut microbiota metabolites
Catalog No. 50-004-35589 Supplier Medchemexpress LLC Supplier No. HY11329110MM/1ML
5'-Deoxyadenosine is an oxidized nucleoside found in the urine of normal subjects. It shows anti-orthopoxvirus activity.
  • Oxidized nucleoside
  • Found in the urine of normal subjects
  • Shows anti-orthopoxvirus activity
  • For research use only
  • Available in solid and solution forms
  • High purity (99.92%)
  • Endogenous metabolite
  • Human gut microbiota metabolites