Learn More
Strem, An Ascensus Company CAS# 894086-00-1. 250MG. 5-(Di-t-butylphosphino)-1',3',5'-triphenyl-1,4'-bi-1H-pyrazole, min. 97% t-Bu-BippyPhos MFCD09038440
SDP
Catalog No. 501385394
Encompass
CAS# 894086-00-1. 250MG. 5-(Di-t-butylphosphino)-1',3',5'-triphenyl-1,4'-bi-1H-pyrazole, min. 97% t-Bu-BippyPhos. MFCD09038440. Molecular Formula: C32H35N4P Molecular Weight: 506.62. Color/Form: white to pale yellow solid. Stability: air sensitive. Strem#: 15-1037 www.strem.com/catalog/v3/15-1037/
Catalog No. 50-138-5394 Supplier Strem, An Ascensus Company Supplier No. 151037250MG
CAS# 894086-00-1. 250MG. 5-(Di-t-butylphosphino)-1',3',5'-triphenyl-1,4'-bi-1H-pyrazole, min. 97% t-Bu-BippyPhos. MFCD09038440. Molecular Formula: C32H35N4P Molecular Weight: 506.62. Color/Form: white to pale yellow solid. Stability: air sensitive. Strem#: 15-1037 www.strem.com/catalog/v3/15-1037/