Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Di-tert-butylphosphino-1',3',5'-triphenyl-1'H-1,4'-bipyrazole, 97%

Catalog No. AAH6386403
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6386403 1 g
AAH63864MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6386403 Supplier Thermo Scientific Chemicals Supplier No. H6386403
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 894086-00-1 | C32H35N4P | 506.634 g/mol

5-(Di-tert-butylphosphino)-1', 3', 5'-triphenyl-1'H-[1,4']bipyrazole is a non-proprietary ligand for palladium-catalyzed amination of aryl halides including aryl chlorides. Works best when the palladium source is Pd2(dba)3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(Di-tert-butylphosphino)-1′, 3′, 5′-triphenyl-1′H-[1,4′]bipyrazole is a non-proprietary ligand for palladium-catalyzed amination of aryl halides including aryl chlorides. Works best when the palladium source is Pd2(dba)3

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 894086-00-1
Molecular Formula C32H35N4P
Molecular Weight (g/mol) 506.634
MDL Number MFCD09038440
InChI Key PTXJGGGNGMPMBG-UHFFFAOYSA-N
Synonym bippyphos, t-bu-bippyphos, 5-di-tert-butylphosphino-1',3',5'-triphenyl-1'h-1,4'-bipyrazole, 5-di-tert-butylphosphino-1', 3', 5'-triphenyl-1'h-1,4' bipyrazole, 5-di-tert-butylphosphino-1',3',5'-triphenyl-1'h-1,4' bipyrazole, 5-di-t-butylphosphino-1',3',5'-triphenyl-1,4'-bi-1h-pyrazole, 5-bis tert-butyl phosphino-1',3',5'-triphenyl-1,4'-bi-1h-pyrazole, 5-di-tert-butylphosphanyl-1',3',5'-triphenyl-1,4'-bipyrazole, 1',3',5'-triphenyl-5-ditert-butylphosphino-1,4'-bi 1h-pyrazole, 5-di-tert-butyl-phosphanyl-1',3',5'-triphenyl-1'h-1,4' bipyrazolyl
PubChem CID 11547931
IUPAC Name ditert-butyl-[2-(1,3,5-triphenylpyrazol-4-yl)pyrazol-3-yl]phosphane
SMILES CC(C)(C)P(C1=CC=NN1C2=C(N(N=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C(C)(C)C

Specifications

Melting Point 169°C to 173°C
Color White
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 506.62
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Di-tert-butylphosphino-1',3',5'-triphenyl-1'H-1,4'-bipyrazole
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.