Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Diethylamino-1-pentanol 98.0+%, TCI America™
SDP

Catalog No. D48651ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48651ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D48651ML Supplier TCI America Supplier No. D48651ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2683-57-0
Molecular Formula C9H21NO
Molecular Weight (g/mol) 159.27
MDL Number MFCD00046024
InChI Key RUQDFMATAGUGMU-UHFFFAOYSA-N
PubChem CID 520284
IUPAC Name 5-(diethylamino)pentan-1-ol
SMILES CCN(CC)CCCCCO

Specifications

Color Yellow
Boiling Point 217°C
Quantity 1 mL
Formula Weight 159.27
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Diethylamino-1-pentanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.