Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Difluoromethoxy)-2-mercaptobenzimidazole 98.0+%, TCI America™
SDP

Catalog No. D343825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D343825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D343825G Supplier TCI America Supplier No. D343825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 97963-62-7
Molecular Formula C8H6F2N2OS
Molecular Weight (g/mol) 216.206
MDL Number MFCD00467504
InChI Key HJMVPNAZPFZXCP-UHFFFAOYSA-N
Synonym 5-difluoromethoxy-2-mercapto-1h-benzimidazole, 5-difluoromethoxy-1h-benzo d imidazole-2-thiol, 5-difluoromethoxy-1h-benzimidazole-2-thiol, unii-39p59c89nv, 5-difluoromethoxy-2-benzimidazolethiol, 5-difluoromethoxy-2-mercaptobenzimidazole, 5-difluoromethoxy-2-thio-benzimidazole, 5-difluoromethoxy-1h-1,3-benzodiazole-2-thiol, 5-difluoromethoxy-1,3-dihydrobenzimidazole-2-thione
PubChem CID 5064774
IUPAC Name 5-(difluoromethoxy)-1,3-dihydrobenzimidazole-2-thione
SMILES C1=CC2=C(C=C1OC(F)F)NC(=S)N2

Specifications

Melting Point 243°C
Color White-Yellow
Quantity 25 g
Formula Weight 216.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(Difluoromethoxy)-2-mercaptobenzimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.