Learn More
Cayman Chemical 5β-Dihydroprogesterone, Purity: ≥95%, CAS: 128-23-4, 500 mg
SDP
Catalog No. 502950314
Encompass_Preferred
A progesterone receptor agonist and metabolite of progesterone; inhibits spontaneous contractions in isolated rat uterus at 10 µg/ml; a negative modulator of homooligomeric ρ1 subunit-containing GABAAreceptors expressed in X. laevis oocytes (IC50 = 5.02 µM); plasma levels decrease at the onset of spontaneous human labor
Catalog No. 50-295-0314 Supplier Cayman Chemical Supplier No. 34644500 MG
A progesterone receptor agonist and metabolite of progesterone; inhibits spontaneous contractions in isolated rat uterus at 10 µg/ml; a negative modulator of homooligomeric ρ1 subunit-containing GABAAreceptors expressed in X. laevis oocytes (IC50 = 5.02 µM); plasma levels decrease at the onset of spontaneous human labor