Learn More
Medchemexpress LLC 5'-DMT-3'-TBDMS-Bz-rA | 81256-88-4 | MFCD00274103 | 99.2% | 787.97 g/mol | C44H49N5O7Si | 1 ML
SDP
Catalog No. 5000301316
Encompass_Preferred
5'-DMT-3'-TBDMS-Bz-rA is a protected adenosine nucleoside reagent used for protection and modification during oligonucleotide synthesis. It is supplied as a ready-to-use 10 mM solution in DMSO and is also offered in solid forms. The product page provides molecular and analytical data plus storage recommendations to support reliable use.
  • Protected adenosine derivative suitable for oligonucleotide synthesis.
  • Supplied as a 10 mM solution in DMSO, ready for use.
  • High purity (99.19%) for consistent analytical performance.
  • Available as solution and solid pack sizes for workflow flexibility.
  • Storage guidance provided for solid and solvent formats to preserve stability.
  • Structural and SMILES data included for compound characterization.
Catalog No. 50-003-01316 Supplier Medchemexpress LLC Supplier No. HYW01817910MM/1ML
5'-DMT-3'-TBDMS-Bz-rA is a protected adenosine nucleoside reagent used for protection and modification during oligonucleotide synthesis. It is supplied as a ready-to-use 10 mM solution in DMSO and is also offered in solid forms. The product page provides molecular and analytical data plus storage recommendations to support reliable use.
  • Protected adenosine derivative suitable for oligonucleotide synthesis.
  • Supplied as a 10 mM solution in DMSO, ready for use.
  • High purity (99.19%) for consistent analytical performance.
  • Available as solution and solid pack sizes for workflow flexibility.
  • Storage guidance provided for solid and solvent formats to preserve stability.
  • Structural and SMILES data included for compound characterization.