Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5'-DMT-3'-TBDMS-Bz-rA | 81256-88-4 | MFCD00274103 | 99.2% | 787.97 g/mol | C44H49N5O7Si | 250 MG
SDP

Catalog No. 5000302384
Encompass_Preferred

5'-DMT-3'-TBDMS-Bz-rA is a protected nucleoside reagent used for nucleoside modification and protection in synthetic applications. It functions as a protected adenosine building block for oligonucleotide assembly and related research, and is supplied with a reported high purity and manufacturer-recommended storage conditions to maintain stability.

  • Protected adenosine derivative for nucleic acid synthesis.
  • Reported high purity (99.2%) for consistent performance.
  • Well-defined molecular properties (C44H49N5O7Si, 787.97 g/mol).
  • Available in multiple lab-scale pack sizes for experimental flexibility.
  • Storage conditions provided to preserve compound stability.

Catalog No. 50-003-02384 Supplier Medchemexpress LLC Supplier No. 250MGHYW018179
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5'-DMT-3'-TBDMS-Bz-rA is a protected nucleoside reagent used for nucleoside modification and protection in synthetic applications. It functions as a protected adenosine building block for oligonucleotide assembly and related research, and is supplied with a reported high purity and manufacturer-recommended storage conditions to maintain stability.

  • Protected adenosine derivative for nucleic acid synthesis.
  • Reported high purity (99.2%) for consistent performance.
  • Well-defined molecular properties (C44H49N5O7Si, 787.97 g/mol).
  • Available in multiple lab-scale pack sizes for experimental flexibility.
  • Storage conditions provided to preserve compound stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.