Learn More
New England Biolabs, Inc. 5’ DNA Adenylation Kit – 50 reactions
SDP
Catalog No. 501526342
Encompass
The 5' DNA adenylation Kit is a simple and efficient enzymatic method for generating 5' -adenylated DNA. The kit is optimized to produce the adenylated DNA with or without 3' -terminator. The 5 DNA adenylation kit routinely generates greater than 95% conversion of pDNA to AppDNA. This highly efficient process eliminates the need for gel isolation of the product and increases overall yield.
Catalog No. 50-152-6342 Supplier New England Biolabs, Inc. Supplier No. E2610L
The 5' DNA adenylation Kit is a simple and efficient enzymatic method for generating 5' -adenylated DNA. The kit is optimized to produce the adenylated DNA with or without 3' -terminator. The 5 DNA adenylation kit routinely generates greater than 95% conversion of pDNA to AppDNA. This highly efficient process eliminates the need for gel isolation of the product and increases overall yield.