Learn More
AdipoGen 5-Dodecanoylaminofluorescein
SDP
Supplier:  AdipoGen CDXD0162M250
Encompass_Preferred
Chemical. CAS 107827-77-0. Formula C32H35NO6. MW 529.62. Synthetic. Lipophilic fluorescent cell staining probe contains a C12 alkyl chain that will bind to membranes with the fluorophore at the aqueous interface and the alkyl tail protruding into the lipid interior. Useful to label plasma membranes. Binds strongly to amphiphilic block copolymer micelles. Useful for determining the critical micelle concentration CMC of various detergents based on fluorescence polarization anisotropy. Lipophilic fluorescein probes are commonly utilized for fluorescence recovery after photobleaching FRAP measurements of lipid lateral diffusion. Used for visualization of proteins in 1-D and 2-D SDS-PAGE gels. Spectral data lambdaex=490nm, lambdaem=520nm.
Catalog No. 50-256-1208