Learn More
AdipoGen 5-DTAF hydrochloride
SDP
Catalog No. 502561226
Encompass_Preferred
Chemical. CAS 21811-74-5. Formula C23H12Cl2N4O5 . HCl. MW 495.3 . 36.5. Synthetic. 5-DTAF is highly reactive with proteins. Unlike other reactive fluoresceins, 5-DTAF not only reacts with amino groups, but also reacts with thiol groups and even directly reacts with hydroxy groups such as polysaccharides and other alcohols in aqueous solution at pH above 9. Due to its high reactivity, 5-DTAF is also used to label carbohydrates. Spectral properties Abs/Em = 492/517 nm
Catalog No. 50-256-1226 Supplier AdipoGen Supplier No. CDXD0123GG25
Chemical. CAS 21811-74-5. Formula C23H12Cl2N4O5 . HCl. MW 495.3 . 36.5. Synthetic. 5-DTAF is highly reactive with proteins. Unlike other reactive fluoresceins, 5-DTAF not only reacts with amino groups, but also reacts with thiol groups and even directly reacts with hydroxy groups such as polysaccharides and other alcohols in aqueous solution at pH above 9. Due to its high reactivity, 5-DTAF is also used to label carbohydrates. Spectral properties Abs/Em = 492/517 nm