Learn More
Roboz Surgical #5 DUMONT INOX TIP .10 X .06MM
SDP
Catalog No. NC9273611
Encompass
Dumont Tweezers; Pattern #5; .10 X .06mm Tip Size; 110mm Length; Material: Inox
Catalog No. NC9273611 Supplier Roboz Surgical Supplier No. RS5045
Dumont Tweezers; Pattern #5; .10 X .06mm Tip Size; 110mm Length; Material: Inox