Learn More
Medchemexpress LLC 5-[(E)-2-thiophen-2-ylethenyl]-1H-pyrazole | 1070881-42-3 | 99.9% | C9H8N2S | 10MG
SDP
Catalog No. 5000258416
Encompass_Preferred
Terevalefim (ANG-3777) is a small-molecule hepatocyte growth factor (HGF) mimetic that selectively activates the c-Met receptor. It has been used in preclinical studies to decrease apoptosis, increase cellular proliferation, and promote organ repair and function. For research use only; not for clinical use or sale to patients.
  • Selectively activates the c-Met receptor in cellular assays.
  • Demonstrated to decrease apoptosis and promote proliferation in preclinical models.
  • High chemical purity suitable for research applications.
  • Solid, white to light brown appearance for convenient handling and storage.
  • Soluble in DMSO at high concentration; ultrasonic agitation recommended.
  • Stable when stored under recommended cryogenic conditions.
Catalog No. 50-002-58416 Supplier Medchemexpress LLC Supplier No. HY13745510MG
Terevalefim (ANG-3777) is a small-molecule hepatocyte growth factor (HGF) mimetic that selectively activates the c-Met receptor. It has been used in preclinical studies to decrease apoptosis, increase cellular proliferation, and promote organ repair and function. For research use only; not for clinical use or sale to patients.
  • Selectively activates the c-Met receptor in cellular assays.
  • Demonstrated to decrease apoptosis and promote proliferation in preclinical models.
  • High chemical purity suitable for research applications.
  • Solid, white to light brown appearance for convenient handling and storage.
  • Soluble in DMSO at high concentration; ultrasonic agitation recommended.
  • Stable when stored under recommended cryogenic conditions.