Learn More
Cayman Chemical 5-EAPBhydrochlorIde 10mg
SDP
Catalog No. 502969639
Encompass_Preferred
An analytical reference standard categorized as a benzofuran; increases conditioned place preference for the 5-EAPB-paired chamber and reinforcement in a self-administration test in rats; has been found in seized samples suspected of containing NPS; associated with fatality; intended for research and forensic applications
Catalog No. 50-296-9639 Supplier Cayman Chemical Supplier No. 1528210 MG
An analytical reference standard categorized as a benzofuran; increases conditioned place preference for the 5-EAPB-paired chamber and reinforcement in a self-administration test in rats; has been found in seized samples suspected of containing NPS; associated with fatality; intended for research and forensic applications