Learn More
Cayman Chemical (±)5-epi CP 55,940, Purity: ≥97%, CAS: 2365471-91-4, 5 mg
SDP
Catalog No. 502955039
Encompass_Preferred
An analytical reference standard that is structurally similar to known synthetic cannabinoids; a potential byproduct in the synthesis of (±)-CP 55,940 based on the published synthesis of (−)-CP 47,497; intended for research and forensic applications
Catalog No. 50-295-5039 Supplier Cayman Chemical Supplier No. 138035 MG
An analytical reference standard that is structurally similar to known synthetic cannabinoids; a potential byproduct in the synthesis of (±)-CP 55,940 based on the published synthesis of (−)-CP 47,497; intended for research and forensic applications