Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Ethoxycarbonyl-2-Fufurylzinc Chloride Solution

Catalog No. 53356450ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-356-450ML 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 53-356-450ML Supplier Sigma Aldrich Supplier No. 53356450ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Product of Rieke™ metals, Inc. ™Registered trademark of Rieke Metals, Inc.

Specifications

CAS 57288-03-6
Melting Point 115.5°C to 119.5°C
Molecular Formula C13H16N2O2
Linear Formula C13H16N2O2
Quantity 500 mg
Synonym [[5-(Ethoxycarbonyl)-2-furanyl]methyl]zinc chloride
Molecular Weight (g/mol) 232.28 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.