Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Ethylpyridine-2,3-dicarboxylic acid, 94%, Thermo Scientific™

Catalog No. AAL1945114
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1945114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1945114 Supplier Thermo Scientific Chemicals Supplier No. L1945114

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102268-15-5
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00071733
InChI Key MTAVBTGOXNGCJR-UHFFFAOYSA-N
Synonym 5-ethylquinolinic acid, 5-ethyl-2,3-pyridinedicarboxylic acid, 2,3-pyridinedicarboxylic acid, 5-ethyl, 5-ethyl-2,3-pyridine dicarboxylic acid, 5-ethylpyridine-2,3-dicarboxylicacid, pubchem16905, dsstox_cid_9072, dsstox_rid_78667, dsstox_gsid_29072, ksc179a3d
PubChem CID 113606
IUPAC Name 5-ethylpyridine-2,3-dicarboxylic acid
SMILES CCC1=CN=C(C(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 152°C to 154°C
Quantity 25 g
Formula Weight 195.18
Percent Purity 94%
Chemical Name or Material 5-Ethylpyridine-2,3-dicarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
RTECSNumber US8005000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.