Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Ethynyl-1,2,3-trifluorobenzene 98.0+%, TCI America™
SDP

Catalog No. E10781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E10781G Supplier TCI America Supplier No. E10781G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 158816-55-8
Molecular Formula C8H3F3
Molecular Weight (g/mol) 156.11
MDL Number MFCD08458177
InChI Key BZXWRVPVZZZAKB-UHFFFAOYSA-N
Synonym 3,4,5-Trifluorophenylacetylene
PubChem CID 22709071
IUPAC Name 5-ethynyl-1,2,3-trifluorobenzene
SMILES FC1=CC(=CC(F)=C1F)C#C

Specifications

Color Yellow
Quantity 1 g
UN Number 1993
Formula Weight 156.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Ethynyl-1,2,3-trifluorobenzene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.