Learn More
Medchemexpress LLC 5'-ethynyl-2'-deoxycytidine | 69075-47-4 | MFCD22417249 | 98.0% | C11H13N3O4 | 10MG
SDP
Catalog No. 5000259262
Encompass_Preferred
5'-Ethynyl-2'-deoxycytidine is a nucleoside analog of deoxycytidine with an ethynyl group at the 5' position. It is used to label newly synthesized DNA via click chemistry and is employed in cell proliferation assays and antiviral research. Supplied as a solid for laboratory research.
  • Click-compatible ethynyl group enables bioorthogonal conjugation.
  • Applicable to DNA synthesis monitoring and cell proliferation assays.
  • Reported activity in antiviral research against HSV.
  • High purity for consistent experimental results.
  • Solid form simplifies storage and handling.
Catalog No. 50-002-59262 Supplier Medchemexpress LLC Supplier No. HY12986110MG
Add to Cart
Add to Cart
5'-Ethynyl-2'-deoxycytidine is a nucleoside analog of deoxycytidine with an ethynyl group at the 5' position. It is used to label newly synthesized DNA via click chemistry and is employed in cell proliferation assays and antiviral research. Supplied as a solid for laboratory research.
  • Click-compatible ethynyl group enables bioorthogonal conjugation.
  • Applicable to DNA synthesis monitoring and cell proliferation assays.
  • Reported activity in antiviral research against HSV.
  • High purity for consistent experimental results.
  • Solid form simplifies storage and handling.