Learn More
Medchemexpress LLC HY-118411 250mg , 5-Ethynyl-2'-deoxyuridine CAS:61135-33-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY118411250MG
Encompass_Preferred
HY-118411 250mg 5-Ethynyl-2'-deoxyuridine CAS: 61135-33-9 5-Ethynyl-2'-deoxyuridine (EdU), a thymidine analogue, is incorporated into cellular DNA during DNA replication and the subsequent reaction of EdU with a fluorescent azide in a “Click” reaction. EdU staining is a fast, sensitive and reproducible method to study cell proliferation. 5-Ethynyl-2'-deoxyuridine is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9643