Learn More
Cayman Chemical 5-Ethynyl-2'-deoxyuridine, Purity: ≥98%, CAS: 61135-33-9, 500 mg
SDP
Catalog No. 502984365
Encompass_Preferred
A nucleoside analog of thymidine that is used to monitor de novo DNA synthesis through click chemistry; can be detected using a fluorescent azide probe and copper catalysis using 1,3-dipolar cycloaddition
Catalog No. 50-298-4365 Supplier Cayman Chemical Supplier No. 20518500 MG
A nucleoside analog of thymidine that is used to monitor de novo DNA synthesis through click chemistry; can be detected using a fluorescent azide probe and copper catalysis using 1,3-dipolar cycloaddition