Learn More
Cayman Chemical 5-Ethynyl-2-deoxyurIdIn 50mg
SDP
Catalog No. 502983614
Encompass_Preferred
A nucleoside analog of thymidine that is used to monitor de novo DNA synthesis through click chemistry; can be detected using a fluorescent azide probe and copper catalysis using 1,3-dipolar cycloaddition
Catalog No. 50-298-3614 Supplier Cayman Chemical Supplier No. 2051850 MG
A nucleoside analog of thymidine that is used to monitor de novo DNA synthesis through click chemistry; can be detected using a fluorescent azide probe and copper catalysis using 1,3-dipolar cycloaddition