Learn More
Medchemexpress LLC HY-118411 50mg Medchemexpress, 5-Ethynyl-2'-deoxyuridine CAS:61135-33-9 Purity:>98%
SDP
Catalog No. 502029841
Encompass_Preferred
Medchemexpress, HY-118411 50mg 5-Ethynyl-2'-deoxyuridine CAS:61135-33-9 5-Ethynyl-2'-deoxyuridine is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9841 Supplier Medchemexpress LLC Supplier No. HY11841150MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-118411 50mg 5-Ethynyl-2'-deoxyuridine CAS:61135-33-9 5-Ethynyl-2'-deoxyuridine is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.