Learn More
Abcam EdU, DNA synthesis monitoring probe, 50MG
Supplier:  Abcam AB14618650MG
Encompass_Preferred
MW 252.22 Da, Purity >98%. Thymidine analog. Alternative to BrdU (5-Bromo-2'-deoxyuridine) (ab142567) for measuring de novo DNA synthesis during S-phase. Cytotoxic anti-metabolite. Used for probing short-term DNA synthesis in tissue culture and living organisms where prolonged cell survival is not required.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1012654