Learn More
Medchemexpress LLC 5-ethynyl-2'-deoxyuridine | 61135-33-9 | 99.8% | 252.23 g/mol | C11H12N2O5 | 1 G
SDP
Catalog No. 5000227615
Encompass_Preferred
5-Ethynyl-2'-deoxyuridine is a thymidine nucleoside analog incorporated into DNA during active DNA synthesis and used to label proliferating cells for detection and imaging via click chemistry.
  • Incorporates into newly synthesized DNA during S-phase.
  • Detectable by click chemistry for sensitive labeling and imaging.
  • Suitable for cell proliferation assays, flow cytometry, and microscopy.
  • High purity solid with defined solubility and storage recommendations for reliable results.
Catalog No. 50-002-27615 Supplier Medchemexpress LLC Supplier No. HY1184111G
5-Ethynyl-2'-deoxyuridine is a thymidine nucleoside analog incorporated into DNA during active DNA synthesis and used to label proliferating cells for detection and imaging via click chemistry.
  • Incorporates into newly synthesized DNA during S-phase.
  • Detectable by click chemistry for sensitive labeling and imaging.
  • Suitable for cell proliferation assays, flow cytometry, and microscopy.
  • High purity solid with defined solubility and storage recommendations for reliable results.