Learn More
Cayman Chemical 5-EthynylurIdIn 100mg
SDP
Catalog No. 502946964
Encompass_Preferred
A clickable form of uridine; is incorporated into cellular RNA and has been used to detect transcription in vitro and in vivo
Catalog No. 50-294-6964 Supplier Cayman Chemical Supplier No. 34391100 MG
A clickable form of uridine; is incorporated into cellular RNA and has been used to detect transcription in vitro and in vivo