Learn More
Cayman Chemical 5-Ethynyluridine, Purity: ≥98%, CAS: 69075-42-9, 10 mg
SDP
Catalog No. 502947685
Encompass_Preferred
A clickable form of uridine; is incorporated into cellular RNA and has been used to detect transcription in vitro and in vivo
Catalog No. 50-294-7685 Supplier Cayman Chemical Supplier No. 3439110 MG
A clickable form of uridine; is incorporated into cellular RNA and has been used to detect transcription in vitro and in vivo